Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770223.1 | XP_008768445.1 | 11 | 1686 | ||
GenBank mRNA | FQ222654.1 | - | 11 | 1620 |