Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam53a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107228.2 | NP_001100698.1 | 3 | 741 | ||
XM_006251262.2 | XP_006251324.1 | 3 | 802 | |||
XM_006251266.2 | XP_006251328.1 | 2 | 703 | |||
XM_006251267.2 | XP_006251329.1 | 2 | 751 | |||
XM_006251268.2 | XP_006251330.1 | 2 | 676 | |||
XM_006251270.2 | XP_006251332.1 | 2 | 216 | |||
XM_008770286.1 | XP_008768508.1 | 2 | 1189 | |||
XM_008770287.1 | XP_008768509.1 | 2 | 1193 | |||
XM_008770288.1 | XP_008768510.1 | 2 | 220 | |||
GenBank mRNA | BC168895.1 | - | 3 | 741 | 7312953 |