Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252370.2 | XP_006252432.1 | 1702 |
Gene Symbol: Kbtbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012045.2 | NP_001012045.2 | 1 | 1440 | ||
GenBank mRNA | BC086326.1 | - | 1279 | 7114652 |