Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1562289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_576917.5 | XP_576917.1 | 1 | 1160 |
Gene Symbol: LOC100360061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729626.3 | XP_002729672.1 | 1079 |
Gene Symbol: LOC100363321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252792.1 | XP_006252854.1 | 1 | 1091 |
Gene Symbol: LOC100909786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750168.3 | XP_003750216.1 | 1 | 1079 |
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013135.2 | NP_001013153.2 | 10 | 1326 | ||
XM_006233681.2 | XP_006233743.1 | 9 | 1236 | |||
XM_006233682.2 | XP_006233744.1 | 9 | 1155 | |||
XM_006233683.2 | XP_006233745.1 | 10 | 1215 | |||
XM_006233684.2 | XP_006233746.1 | 8 | 1068 | |||
XM_006233685.2 | XP_006233747.1 | 8 | 1092 | |||
XM_006233686.2 | XP_006233748.1 | 7 | 1056 | |||
GenBank mRNA | BC088296.1 | - | 4 | 266 | 7322728 | |
BC168170.1 | - | 5 | 2613 | 9025284 |