Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP9X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039590.2 | NP_001034679.2 | 33 | 5669 | ||
NM_001039591.2 | NP_001034680.2 | 33 | 5669 | |||
XM_005272675.4 | XP_005272732.1 | 33 | 5899 | |||
XM_005272676.4 | XP_005272733.1 | 33 | 5853 | |||
GenBank mRNA | AB209666.1 | - | 4 | 656 | ||
BC063645.1 | - | 2 | 130 | |||
X98296.1 | - | 32 | 5074 |