Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfhas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107316.1 | NP_001100786.1 | 1 | 1095 | ||
XM_006253224.2 | XP_006253286.1 | 1 | 1178 | |||
XM_006253225.2 | XP_006253287.1 | 1 | 1178 | |||
XM_006253227.2 | XP_006253289.1 | 1 | 1178 |