Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107322.2 | NP_001100792.1 | 3 | 203 | ||
XM_008771356.1 | XP_008769578.1 | 3 | 393 | |||
XM_008771357.1 | XP_008769579.1 | 3 | 392 | |||
XM_008771358.1 | XP_008769580.1 | 3 | 1533 | |||
XM_008771359.1 | XP_008769581.1 | 3 | 360 |