Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003492.2 | NP_003483.1 | 2 | 1366 | ||
XM_011531199.2 | XP_011529501.1 | 2 | 1381 | |||
GenBank mRNA | BC008203.1 | - | 2 | 1114 | 3868838 | |
BC017026.1 | - | 2 | 1101 | 3847234 | ||
X92475.1 | - | 2 | 1116 |