Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254738.1 | NP_001241667.1 | 5 | 821 | ||
NM_005168.4 | NP_005159.1 | 6 | 728 | |||
GenBank mRNA | AF498969.1 | - | 5 | 543 | ||
AK299731.1 | - | 5 | 504 | |||
AK309442.1 | - | 4 | 402 | |||
AK311312.1 | - | 2 | 295 | |||
AK313224.1 | - | 5 | 795 | |||
BC012513.1 | - | 6 | 683 | 4453777 | ||
BQ440721.1 | - | 5 | 797 | |||
BT006769.1 | - | 5 | 543 | |||
CR542038.1 | - | 5 | 498 | |||
CR542070.1 | - | 5 | 498 | |||
S82240.1 | - | 5 | 522 | |||
X95282.1 | - | 5 | 543 | |||
X97758.1 | - | 5 | 765 |