Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_225268.5 | XP_225268.3 | 3 | 384 |
Gene Symbol: LOC102553010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771592.1 | XP_008769814.1 | 1 | 100 |