Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107358.1 | NP_001100828.1 | 4 | 1939 | ||
XM_006254021.2 | XP_006254083.1 | 4 | 2175 | |||
XM_006254023.2 | XP_006254085.1 | 4 | 1937 | |||
XM_006254027.2 | XP_006254089.1 | 3 | 1785 | |||
XM_008771729.1 | XP_008769951.1 | 5 | 3378 | |||
XM_008771730.1 | XP_008769952.1 | 3 | 2401 |