Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PICALM SIRNA Assays ›
Gene Symbol:
PICALM
Gene Name:
phosphatidylinositol binding clathrin assembly protein
Gene Aliases:
CALM, CLTH, LAP
Chromosome Location:
Chr.11: 85957171 - 86069881 on Build GRCh38
UniGene ID:
Hs.163893
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s15800
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PICALM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008660.2
NP_001008660.1
10
1252
NM_001206946.1
NP_001193875.1
10
1252
NM_001206947.1
NP_001193876.1
10
967
NM_007166.3
NP_009097.2
10
1252
XM_005274322.2
XP_005274379.1
10
1236
XM_005274323.2
XP_005274380.1
10
1236
XM_005274324.2
XP_005274381.1
10
1236
XM_005274325.2
XP_005274382.1
10
1236
XM_005274326.2
XP_005274383.1
10
1236
XM_005274327.2
XP_005274384.1
10
1236
XM_005274328.2
XP_005274385.1
10
1236
XM_005274329.3
XP_005274386.1
10
1236
XM_005274330.2
XP_005274387.1
10
1236
XM_005274331.2
XP_005274388.1
10
1236
XM_005274332.2
XP_005274389.1
10
1236
XM_005274333.2
XP_005274390.1
10
1236
XM_005274334.2
XP_005274391.1
10
1236
XM_005274335.2
XP_005274392.1
10
1236
XM_005274336.2
XP_005274393.1
10
1236
XM_005274337.2
XP_005274394.1
10
1236
XM_005274340.2
XP_005274397.1
10
1236
XM_006718699.2
XP_006718762.1
10
1236
XM_006718700.2
XP_006718763.1
10
1236
XM_006718701.3
XP_006718764.1
10
1236
XM_011545293.1
XP_011543595.1
10
1236
XM_017018381.1
XP_016873870.1
10
1236
XM_017018382.1
XP_016873871.1
10
1236
XM_017018383.1
XP_016873872.1
10
1236
XM_017018384.1
XP_016873873.1
10
1236
XM_017018385.1
XP_016873874.1
10
1236
XM_017018386.1
XP_016873875.1
10
1236
XM_017018387.1
XP_016873876.1
10
1236
XM_017018388.1
XP_016873877.1
10
1236
GenBank mRNA
AB210017.1
-
10
1252
AB451308.1
-
10
935
AK291414.1
-
11
1119
AK297062.1
-
2
137
AK300275.1
-
10
944
BC048259.1
-
10
1013
5537605
BC064357.1
-
10
1214
5112878
BC073961.1
-
10
1088
6183858
U45976.1
-
10
1082
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
receptor-mediated endocytosis
cell proliferation
negative regulation of gene expression
synaptic vesicle maturation
vesicle-mediated transport
endosomal transport
regulation of endocytosis
receptor internalization
regulation of protein localization
cargo loading into vesicle
positive regulation of transcription, DNA-templated
negative regulation of receptor-mediated endocytosis
clathrin coat assembly
iron ion homeostasis
clathrin-mediated endocytosis
iron ion import into cell
cell-cell adhesion
positive regulation of neuron death
positive regulation of beta-amyloid formation
regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
positive regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
protein binding
1-phosphatidylinositol binding
clathrin binding
clathrin heavy chain binding
clathrin adaptor activity
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f2nf4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0