Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccdc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772250.1 | XP_008770472.1 | 9 | 1002 | ||
XM_008772251.1 | XP_008770473.1 | 9 | 923 | |||
XM_008772252.1 | XP_008770474.1 | 9 | 1210 | |||
XM_008772253.1 | XP_008770475.1 | 8 | 1081 | |||
XM_008772254.1 | XP_008770476.1 | 9 | 1213 | |||
XM_008772255.1 | XP_008770477.1 | 8 | 1060 | |||
XM_008772256.1 | XP_008770478.1 | 8 | 1057 | |||
XM_008772257.1 | XP_008770479.1 | 9 | 1213 |