Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178010.2 | NP_001171481.1 | 3 | 330 | ||
NM_001178011.2 | NP_001171482.1 | 3 | 330 | |||
NM_003504.4 | NP_003495.1 | 3 | 330 | |||
XM_005261285.2 | XP_005261342.1 | 4 | 423 | |||
XM_005261286.3 | XP_005261343.1 | 4 | 423 | |||
XM_011530415.1 | XP_011528717.1 | 4 | 423 | |||
XM_011530416.1 | XP_011528718.1 | 3 | 226 | |||
XM_011530417.2 | XP_011528719.1 | 4 | 624 | |||
XM_011530418.2 | XP_011528720.1 | 4 | 624 | |||
GenBank mRNA | AF053074.1 | - | 3 | 211 | ||
AF062495.1 | - | 3 | 203 | |||
AF081535.1 | - | 3 | 218 | |||
AJ223728.1 | - | 3 | 217 | |||
AK293123.1 | - | 3 | 265 | |||
AK293338.1 | - | 3 | 227 | |||
AK302590.1 | - | 3 | 204 | |||
AK303274.1 | - | 3 | 250 | |||
AY358971.1 | - | 3 | 253 | |||
BC006232.2 | - | 3 | 248 | 3946493 | ||
BC010022.2 | - | 3 | 233 | 2964592 | ||
BM751026.1 | - | 3 | 265 | |||
BT006792.1 | - | 3 | 187 | |||
CT841513.1 | - | 3 | 191 | |||
HY016491.1 | - | 4 | 475 |