Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107428.2 | NP_001100898.2 | 551 | |||
XM_006255588.2 | XP_006255650.1 | 725 | ||||
XM_006255589.2 | XP_006255651.1 | 602 | ||||
XM_006255590.2 | XP_006255652.1 | 3, 4 | 3604 | |||
XM_008772509.1 | XP_008770731.1 | 2594 | ||||
XM_008772510.1 | XP_008770732.1 | 2, 3 | 4198 | |||
XM_008772511.1 | XP_008770733.1 | 2, 3 | 623 |