Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107439.1 | NP_001100909.1 | 3 | 772 | ||
XM_006255765.2 | XP_006255827.1 | 3 | 1182 | |||
XM_008772611.1 | XP_008770833.1 | 3 | 4195 | |||
XM_008772612.1 | XP_008770834.1 | 4 | 5439 |