Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Acat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006995.1 | NP_001006996.1 | 9 | 1116 | ||
GenBank mRNA | AY325187.1 | - | 1813 | |||
BC083872.1 | - | 9 | 1116 | 7128564 |
Gene Symbol: RGD1562948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748697.3 | XP_003748745.2 | 9 | 1210 |