Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100845.2 | NP_001094315.2 | 18 | 4054 | ||
XM_008759140.1 | XP_008757362.1 | 18 | 4095 | |||
XM_008759141.1 | XP_008757363.1 | 18 | 4095 |