Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030055.1 | NP_001025226.1 | 6 | 4470 | ||
NM_001173.2 | NP_001164.2 | 6 | 4467 | |||
XM_005267635.3 | XP_005267692.1 | 6 | 4439 | |||
XM_005267636.3 | XP_005267693.1 | 6 | 4685 | |||
XM_017021287.1 | XP_016876776.1 | 6 | 4389 | |||
XM_017021288.1 | XP_016876777.1 | 6 | 4436 | |||
GenBank mRNA | AB209751.1 | - | 5 | 3065 | ||
AK307454.1 | - | 4 | 324 | |||
AL045112.1 | - | 3 | 241 | |||
BC075799.1 | - | 4 | 652 | |||
BC129928.1 | - | 5 | 562 | |||
BC129929.1 | - | 4 | 417 | |||
U17032.1 | - | 6 | 4448 |