Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107520.1 | NP_001100990.1 | 47 | 7682 | ||
XM_006229300.2 | XP_006229362.1 | 48 | 7870 | |||
XM_006229301.2 | XP_006229363.1 | 48 | 7898 | |||
XM_006229302.2 | XP_006229364.1 | 48 | 7855 | |||
XM_006229304.2 | XP_006229366.1 | 48 | 7870 |