Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034147.1 | NP_001029319.1 | 8, 9 | 841 | ||
GenBank mRNA | BC100091.1 | - | 8, 9 | 841 | 7371990 | |
FQ213073.1 | - | 8, 9 | 887 |