Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034147.1 | NP_001029319.1 | 5 | 507 | ||
GenBank mRNA | BC100091.1 | - | 5 | 507 | 7371990 | |
FQ213073.1 | - | 5 | 553 |