Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107529.1 | NP_001100999.1 | 6, 7 | 941 | ||
XM_006229433.2 | XP_006229495.1 | 6, 7 | 953 | |||
XM_006229435.2 | XP_006229497.1 | 6, 7 | 953 | |||
XM_006229437.2 | XP_006229499.1 | 6, 7 | 953 |