Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006997.1 | NP_001006998.1 | 3, 4 | 1344 | ||
GenBank mRNA | BC083780.1 | - | 3, 4 | 1344 | 7096039 |