Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pign | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100584.1 | NP_001094054.1 | 10 | 1275 | ||
XM_008769468.1 | XP_008767690.1 | 10 | 1322 | |||
XM_008769469.1 | XP_008767691.1 | 10 | 1260 | |||
XM_008769470.1 | XP_008767692.1 | 10 | 1567 | |||
XM_008769471.1 | XP_008767693.1 | 9 | 1239 | |||
XM_008769472.1 | XP_008767694.1 | 9 | 1484 | |||
XM_008769473.1 | XP_008767695.1 | 9 | 1206 | |||
XM_008769474.1 | XP_008767696.1 | 9 | 1177 | |||
XM_008769475.1 | XP_008767697.1 | 8 | 970 | |||
XM_008769476.1 | XP_008767698.1 | 8 | 1123 | |||
XM_008769477.1 | XP_008767699.1 | 8 | 1368 | |||
XM_008769478.1 | XP_008767700.1 | 10 | 1323 |