Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165417.1 | NP_001158889.1 | 2 | 401 | ||
NM_003562.4 | NP_003553.2 | 2 | 434 | |||
GenBank mRNA | AF070548.1 | - | 2 | 288 | ||
AK290376.1 | - | 2 | 185 | |||
BC006508.2 | - | 2 | 290 | 2960933 | ||
BC006519.2 | - | 2 | 290 | 2960933 | ||
BC016294.2 | - | 2 | 209 | 3911858 | ||
BC017170.2 | - | 2 | 290 | 2960933 | ||
BM047391.1 | - | 2 | 148 | |||
CB155476.1 | - | 2 | 434 | |||
CR456834.1 | - | 2 | 177 |