Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tigd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107573.1 | NP_001101043.1 | 2 | 403 | ||
XM_006230805.2 | XP_006230867.1 | 2 | 1157 |