Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Batf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230953.2 | XP_006231015.1 | 4 | 402 | ||
XM_008760203.1 | XP_008758425.1 | 3 | 987 | |||
XM_008760204.1 | XP_008758426.1 | 3 | 986 | |||
XM_219527.7 | XP_219527.5 | 3 | 331 |