Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gna14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013151.1 | NP_001013169.1 | 2 | 711 | ||
XM_008760288.1 | XP_008758510.1 | 3 | 1348 | |||
GenBank mRNA | BC090316.1 | - | 2 | 711 | 7305112 |