Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107589.1 | NP_001101059.1 | 6, 7 | 645 | ||
GenBank mRNA | FQ216123.1 | - | 6, 7 | 655 | ||
FQ216298.1 | - | 6, 7 | 655 | |||
FQ216609.1 | - | 6, 7 | 657 | |||
FQ216805.1 | - | 6, 7 | 655 |