Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261408.1 | NP_001248337.1 | 10 | 1991 | ||
NM_001261409.1 | NP_001248338.1 | 8 | 1977 | |||
NM_001261410.1 | NP_001248339.1 | 6 | 1644 | |||
NM_001308251.1 | NP_001295180.1 | 4 | 964 | |||
NM_003567.3 | NP_003558.1 | 8 | 1954 | |||
XM_011542251.2 | XP_011540553.1 | 7 | 1593 | |||
XM_017002480.1 | XP_016857969.1 | 10 | 2367 | |||
GenBank mRNA | AF124250.1 | - | 8 | 1946 | ||
AK027785.1 | - | 4 | 1212 | |||
AK055442.1 | - | 8 | 1977 | |||
AK301570.1 | - | 4 | 937 | |||
AK310970.1 | - | 4 | 964 | |||
AL833121.1 | - | 6 | 1597 | |||
AY358996.1 | - | 4 | 884 | |||
BC028477.1 | - | 10 | 2020 | |||
BC039895.1 | - | 8 | 1847 | 6060541 | ||
U92715.1 | - | 8 | 1762 |