Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ring1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212549.2 | NP_997714.2 | 4 | 467 | ||
GenBank mRNA | AB568263.1 | - | 3 | 251 | ||
BC098635.1 | - | 4 | 411 | 7098264 |