Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277057.1 | NP_001263986.1 | 13 | 2342 | ||
XM_006256602.2 | XP_006256664.1 | 14 | 2440 | |||
XM_006256603.2 | XP_006256665.1 | 15 | 2548 | |||
XM_008772992.1 | XP_008771214.1 | 16 | 2617 | |||
XM_008772993.1 | XP_008771215.1 | 14 | 2461 |