Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107644.3 | NP_001101114.1 | 1 | 605 | ||
XM_006256589.2 | XP_006256651.1 | 1 | 626 | |||
XM_006256590.2 | XP_006256652.1 | 1 | 626 | |||
XM_008772999.1 | XP_008771221.1 | 2 | 294 | |||
GenBank mRNA | BC082054.1 | - | 1 | 626 | ||
BC166465.1 | - | 1 | 609 | 9030981 |