Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005384.1 | NP_001005384.1 | 17 | 2708 | ||
XM_006232012.2 | XP_006232074.1 | 17 | 2726 | |||
GenBank mRNA | AB167522.1 | - | 17 | 2708 |