Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nim1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231982.2 | XP_006232044.1 | 4 | 3572 | ||
XM_008760801.1 | XP_008759023.1 | 4 | 2086 | |||
XM_227081.6 | XP_227081.3 | 4 | 2702 |