Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mki67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271366.1 | NP_001258295.1 | 14 | 9199 | ||
XM_006230453.2 | XP_006230515.1 | 13 | 9560 | |||
GenBank mRNA | FQ227329.1 | - | 2 | 177 | ||
FQ230341.1 | - | 2 | 177 |
Gene Symbol: LOC100910502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771840.1 | XP_008770062.1 | 2 | 1093 |
Gene Symbol: LOC100912428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770132.1 | XP_008768354.1 | 2 | 1123 |