Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047886.1 | NP_001041351.1 | 8 | 704 | ||
XM_006232624.2 | XP_006232686.1 | 6 | 493 | |||
GenBank mRNA | AY325142.1 | - | 8 | 704 | ||
BC167105.1 | - | 6 | 393 | 9102039 |