Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257197.1 | NP_001244126.1 | 5 | 1035 | ||
NM_001257198.1 | NP_001244127.1 | 6 | 916 | |||
NM_003590.4 | NP_003581.1 | 6 | 1233 | |||
XM_006712800.2 | XP_006712863.2 | 6 | 906 | |||
XM_011511994.2 | XP_011510296.1 | 5 | 790 | |||
XM_011511995.1 | XP_011510297.1 | 6 | 818 | |||
XM_011511996.2 | XP_011510298.1 | 6 | 882 | |||
GenBank mRNA | AB014517.2 | - | 6 | 1161 | ||
AF062537.1 | - | 6 | 934 | |||
AF064087.1 | - | 6 | 926 | |||
AK291151.1 | - | 6 | 883 | |||
AK299644.1 | - | 5 | 645 | |||
AK308496.1 | - | 6 | 916 | |||
AY337761.1 | - | 6 | 1379 | |||
BC039598.1 | - | 6 | 1178 | 5784147 | ||
BC092409.1 | - | 6 | 1178 | 6168545 | ||
U58089.1 | - | 2 | 272 |