Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107712.2 | NP_001101182.2 | 4 | 258 | ||
XM_006233087.1 | XP_006233149.1 | 3 | 235 | |||
GenBank mRNA | CB724536.1 | - | 4 | 276 |