Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127485.1 | NP_001120957.1 | 2 | 1136 | ||
XM_008761400.1 | XP_008759622.1 | 2 | 970 |