Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107722.1 | NP_001101192.1 | 9 | 2255 | ||
XM_006233270.2 | XP_006233332.1 | 7 | 2652 |