Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107726.1 | NP_001101196.1 | 3 | 306 | ||
XM_006232003.1 | XP_006232065.1 | 5 | 1510 |
Set Membership: |
![]() |