Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEB SIRNA Assays ›
Gene Symbol:
Neb
Gene Name:
nebulin
Gene Aliases:
-
Chromosome Location:
Chr.3: 37658081 - 37855834 on Build Rnor_6.0
UniGene ID:
Rn.134031
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s160832
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Neb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_006234174.2
XP_006234236.1
30, 31
3234
XM_006234175.2
XP_006234237.1
30, 31
3234
XM_006234176.2
XP_006234238.1
30, 31
3234
XM_006234177.2
XP_006234239.1
30, 31
3234
XM_006234178.2
XP_006234240.1
30, 31
3234
XM_006234179.2
XP_006234241.1
30, 31
3234
XM_006234180.2
XP_006234242.1
30, 31
3234
XM_006234181.2
XP_006234243.1
30, 31
3234
XM_006234182.2
XP_006234244.1
30, 31
3234
XM_006234183.2
XP_006234245.1
30, 31
3234
XM_006234185.2
XP_006234247.1
30, 31
3234
XM_006234186.2
XP_006234248.1
30, 31
3234
XM_006234188.2
XP_006234250.1
30, 31
3234
XM_006234190.2
XP_006234252.1
30, 31
3234
XM_006234191.2
XP_006234253.1
30, 31
3234
XM_006234192.2
XP_006234254.1
30, 31
3234
XM_006234193.2
XP_006234255.1
30, 31
3234
XM_006234194.2
XP_006234256.1
30, 31
3234
XM_008761815.1
XP_008760037.1
30, 31
3235
XM_008761816.1
XP_008760038.1
30, 31
3234
XM_008761817.1
XP_008760039.1
30, 31
3234
XM_008761818.1
XP_008760040.1
30, 31
3234
XM_008761819.1
XP_008760041.1
30, 31
3234
XM_008761820.1
XP_008760042.1
30, 31
3235
XM_008761821.1
XP_008760043.1
30, 31
3235
Back To Top
More Information
Gene Ontology Categories:
Process(es)
actin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-nglqg:80/10.239.166.193:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0