Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127486.1 | NP_001120958.1 | 4 | 728 | ||
XM_008761895.1 | XP_008760117.1 | 4 | 666 | |||
GenBank mRNA | BC158633.1 | - | 4 | 729 | 7456638 |