Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107746.1 | NP_001101216.1 | 4 | 1124 | ||
XM_006234507.2 | XP_006234569.1 | 4 | 1116 | |||
XM_006234508.1 | XP_006234570.1 | 4 | 1206 |