Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762226.1 | XP_008760448.1 | 13 | 1252 | ||
XM_008762227.1 | XP_008760449.1 | 13 | 1603 | |||
XM_008762228.1 | XP_008760450.1 | 12 | 2224 | |||
XM_008762229.1 | XP_008760451.1 | 12 | 1486 | |||
XM_008762230.1 | XP_008760452.1 | 12 | 1138 |