Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ap4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191742.1 | NP_001178671.1 | 8 | 919 | ||
XM_006234921.1 | XP_006234983.1 | 5 | 565 | |||
XM_008762176.1 | XP_008760398.1 | 4 | 1379 |