Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIBP1 SIRNA Assays ›
Gene Symbol:
PPFIBP1
Gene Name:
PPFIA binding protein 1
Gene Aliases:
L2, SGT2, hSGT2, hSgt2p
Chromosome Location:
Chr.12: 27524112 - 27695564 on Build GRCh38
UniGene ID:
Hs.172445
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16134
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198916.1
NP_001185845.1
4
457
NM_003622.3
NP_003613.3
4
457
NM_177444.2
NP_803193.2
4
457
XM_005253505.1
XP_005253562.1
3
160
XM_005253507.1
XP_005253564.1
3
160
XM_005253508.1
XP_005253565.1
3
160
XM_005253509.1
XP_005253566.1
3
160
XM_017020056.1
XP_016875545.1
5
476
XM_017020057.1
XP_016875546.1
3
651
XM_017020058.1
XP_016875547.1
3
160
XM_017020059.1
XP_016875548.1
4
443
XM_017020060.1
XP_016875549.1
3
160
XM_017020061.1
XP_016875550.1
4
426
XM_017020062.1
XP_016875551.1
4
440
XM_017020063.1
XP_016875552.1
4
443
XM_017020064.1
XP_016875553.1
4
426
XM_017020065.1
XP_016875554.1
4
426
XM_017020066.1
XP_016875555.1
4
426
XM_017020067.1
XP_016875556.1
4
426
XM_017020068.1
XP_016875557.1
4
442
XM_017020069.1
XP_016875558.1
4
426
XM_017020070.1
XP_016875559.1
4
426
XM_017020071.1
XP_016875560.1
4
426
XM_017020072.1
XP_016875561.1
4
426
XM_017020073.1
XP_016875562.1
4
443
XM_017020077.1
XP_016875566.1
3
160
XM_017020078.1
XP_016875567.1
4
431
GenBank mRNA
AB049149.1
-
4
349
AF034802.1
-
4
320
AJ276680.1
-
3
188
AK314350.1
-
4
345
BC022873.1
-
4
428
BC046159.1
-
4
274
BC050281.1
-
4
363
5276255
BM272389.1
-
4
460
BQ777042.1
-
4
467
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
cell-cell adhesion
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0