Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPFIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304331.1 | NP_001291260.1 | 26 | 3602 | ||
NM_001304332.1 | NP_001291261.1 | 25 | 3575 | |||
XM_006711586.2 | XP_006711649.1 | 26 | 3632 | |||
XM_011510070.2 | XP_011508372.1 | 14 | 1661 | |||
XM_017002606.1 | XP_016858095.1 | 25 | 3605 | |||
XM_017002607.1 | XP_016858096.1 | 25 | 3497 | |||
XM_017002608.1 | XP_016858097.1 | 25 | 3605 | |||
XM_017002609.1 | XP_016858098.1 | 25 | 3497 | |||
GenBank mRNA | AB020704.1 | - | 8 | 731 | ||
AF034801.1 | - | 10 | 1126 | |||
AK023365.1 | - | 7 | 772 | |||
AK124535.1 | - | 2 | 1089 | |||
AK126624.1 | - | 13 | 2170 | |||
BC132923.1 | - | 13 | 2112 | 40146945 | ||
BC132925.1 | - | 13 | 2112 | 40146947 | ||
BC144262.1 | - | 12 | 2085 | 9052782 | ||
EF428334.1 | - | 31 | 3469 |